Calcineurin Cellular Activity Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK816-0001
89156-556EA 716.13 USD
89156-556
Calcineurin Cellular Activity Assay Kit, Enzo Life Sciences
Assays Cellular Assays
A complete colorimetric assay kit for measuring cellular calcineurin (PP2B) phosphatase activity
The RII phosphopeptide substrate, supplied with this kit is the most efficient and selective peptide known for calcineurin. The detection of free-phosphate released is based on the classic malachite green assay.
It employs a convenient 96-well plate format with all reagents necessary for measuring calcineurin (PP2B) phosphatase activity in tissue/cellular extracts PLUS human recombinant calcineurin is included as a positive control.
Delivery information: Kit includes Calcineurin enzyme, Calmodulin (human, recombinant), Substrate (RII phosphopeptide), 2x assay buffer, EGTA Buffer, Lysis Buffer, Protease inhibitor cocktail, Biomol Green™ Reagent, Phosphate standard, Okadaic acid, Desalting column and resin, Half Volume microplate.