Caspase-1 Assay Kit for Drug Discovery, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK701-0001
89156-536EA 794.61 USD
89156-536
Caspase-1 Assay Kit for Drug Discovery, Enzo Life Sciences
Assays Protein Assays
This kit is a complete assay system to measure protease activity of recombinant caspase-1
Cleavage of a tetrapeptide substrate is monitored colorimetrically at 405 nm or fluorimetrically (Ex. 360 nm/Em. 460 nm).
Delivery information: Kit includes Caspase-1 enzyme (human) (recombinant), Ac-YVAD-pNA colorimetric substrate, Ac-YVAD-AMC fluorogenic substrate, calibration standards, aldehyde inhibitor, assay buffer, half volume 96-microwell plate, and instruction booklet.