Cathepsin K Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK430-0001
89156-492EA 709.59 USD
89156-492
Cathepsin K Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
The Cathepsin K Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin K. Using a convenient microplate format, cathepsin K activity is monitored fluorimetrically using the substrate provided in the kit.
Ordering information: Quantity: 96 assays Contents: Cathepsin K (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer, 1/2-volume white microplate.