Matrix Metalloproteinase-14 (MMP-14) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK417-0001
89156-484EA 733.57 USD
89156-484
Matrix Metalloproteinase-14 (MMP-14) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZymeTM Assay System
The MMP-14 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay sytem designed to screen for inhibitors of matrix metalloproteinase-14 (MMP-14, membrane-type matrix metalloproteinase 1, MT1-MMP) using a quenched fluorogenic peptide: OmniMMPTM fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties.
Delivery information: Kit contains MMP-14 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.