Matrix Metalloproteinase-8 (MMP-8) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK414-0001
89156-478EA
727.03
USD
89156-478
Matrix Metalloproteinase-8 (MMP-8) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Assays
Enzyme Assays
A QuantiZymeTM Assay System
The MMP-8 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) using a thiopeptide as a chromogenic substrate.
Delivery information: Kit contains MMP-8 Enzyme (human) (recombinant), Substrate,, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)