Matrix Metalloproteinase-2 (MMP-2) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK409-0001
89156-468EA 733.57 USD
89156-468
Matrix Metalloproteinase-2 (MMP-2) Fluorometric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantizymeTM Assay System
The MMP-2 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-2 (MMP-2, gelatinase A) using a quenched fluorogenic peptide: OmniMMPTM fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties. The assays are performed in a convenient 96-well microplate format. The kit is useful to screen inhibitors of MMP-2, a potential therapeutic target.
Delivery information: Kit contains MMP-2 enzyme (human) (recombinant), substrate, calibration standard, inhibitor, assay buffer and 96-well white microplate.