Matrix Metalloproteinase-7 (MMP-7) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK406-0001
89156-462EA 727.03 USD
89156-462
Matrix Metalloproteinase-7 (MMP-7) Colorimetric Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
A QuantiZymeTM Assay SystemThe MMP-7 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-7 (MMP-7, matrilysin) using a thiopeptide as a chromogenic substrate.
A QuantiZymeTM Assay SystemThe MMP-7 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-7 (MMP-7, matrilysin) using a thiopeptide as a chromogenic substrate The assays are performed in a convenient 96-well microplate format. Included are active enzyme, assay buffer, a prototypic control inhibitor (NNGH), and a detailed instruction booklet.
Delivery information: Kit contains MMP-7 Enzyme (human) (recombinant), Substrate, Prototypic Control Inhibitor (NNGH), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.