Mycophenolic acid ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

BML-A249-0500
89156-352EA 251 USD
89156-352 89156-354
Mycophenolic acid ≥98% (by TLC)
Mycophenolic acid

Inosine-5'-monophosphate dehydrogenase inhibitor


Antiviral and immunosuppressive activity. Clinically useful antipsoriatic agent. Mycophenolic acid prodrugs are effective in rheumatoid arthritis and for the prevention of organ transplant rejection. Mechanism of action is inhibition of inosine-5'-monophosphate dehydrogenase.


Isolated from Penicillium brevi-compactum

Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
MDL Number: MFCD00036814
CAS Number: 24280-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33f00efb38a719dd9eff282783253efc6a19410a&vid=72a95b20397162e7bf113250cbdcb113a0216d49&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR