SCREEN-WELL® Orphan Ligand Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-2825-0500
89156-288EA 3066.17 USD
89156-288
SCREEN-WELL® Orphan Ligand Library, Enzo Life Sciences
Microplates Immunology Microplates and Strips
The SCREEN-WELL® Orphan ligand library contains 84 compounds with defined, putative, potential or speculative biological activity but whose protein binding partners have not been identified.
Ligands are supplied dissolved in biocompatible solvents at 10mM and aliquoted into a deep-well plate at 500µL per well
Stable for one year from the date of receipt when stored at -80°C and shipped on Dry Ice.
The library is a rich source of ligands for receptor de-orphaning. Includes the following classes of ligands: Trace Amines, Neurotransmitter Metabolites, Endogenous β-carbolines, Urinary Metabolites, Nicotine Congeners, D-Amino Acids. Receptors for endogenous trace amines have been deorphanized. Endogenous β-carbolines have been shown to display varied biological effects such as hallucinations, hypotension and others. Nicotine congeners such as nornicotine are biologically active and may act at novel nicotinic receptor subtypes.
Ordering information: Quantity: 84 compounds, 500μl per compound
Delivery information: 84 compounds, 500 µL per compound