Annexin V-EGFP Apoptosis Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-253-KI02
89156-042EA
596.23
USD
89156-042
Annexin V-EGFP Apoptosis Detection Kit, Enzo Life Sciences
Assays
Protein Assays
For the detection of apoptotic cells by flow cytometry.
EGFP (enhanced green fluorescent protein) has emerged as an ideal fluorochrome for FACS analysis and fluorescent microscopy as it offers a more intense signal than wild-type GFP. It has Ex(max): 488 nm and Em(max): 530 nm. The green signal produced makes EGFP ideal for dual staining with propidium iodide (PI).
Delivery information: Kit contains Annexin V-EGFP, propidium iodide solution, binding buffer and assay protocol.