Live-Dead Cell Staining Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-249-KI01
89156-020EA 479.6 USD
89156-020
Live-Dead Cell Staining Kit, Enzo Life Sciences
Assays Cellular Assays
Kit utilizes Live-dye™, a cell permeable green fluorescent dye (Ex(max): 488nm; Em(max): 518nm), to stain live cells. Dead cells can be easily stained by propidium iodide (PI), a red fluorescent dye (Ex(max): 488nm; Em(max): 615nm), which in viable cells is actively pumped out of the cytoplasm.
  • Ready-to-use
  • Live-dead cell stain for 100 tests
Stained cells can be visualized by fluorescence microscopy using a band-pass filter (detects FITC and rhodamine). Cyto-dye has also been used to discriminate between apoptotic and non-apoptotic cells.