Caspase-2 Fluorometric Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-214-KI01
89155-956EA 300.84 USD
89155-956
Caspase-2 Fluorometric Assay Kit, Enzo Life Sciences
Assays Protein Assays
Based on the detection of AFC (7-amino-4-trifluoromethyl coumarin) after cleavage from the labelled substrate VDVAD-AFC.
Delivery information: Kit includes Cell Lysis buffer (25 mL), 2X Reaction Buffer (2 mL), 1 mM VDVAD-AFC Substrate (125 µL), DTT (100 µL).