Annexin V-FITC Apoptosis Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
850-020-KI01
850-020-KI02
89155-876EA
851.29
USD
89155-876
89155-878
Annexin V-FITC Apoptosis Detection Kit, Enzo Life Sciences
Assays
Protein Assays
For the detection of apoptotic cells by flow cytometry.
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488 nm, Em(max): 530 nm).
Delivery information: Kit contains Annexin V-FITC, binding buffer and propidium iodide.