Anti-PTPRC Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
810-410-L015
89155-752EA 610.4 USD
89155-752
Anti-PTPRC Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit, Isotype: IgG

  • For Immunohistochemistry

Recognizes human CD45.

Type: Primary
Antigen: PTPRC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR