Anti-ITGB2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
805-062-C100
89155-336EA 397.85 USD
89155-336
Anti-ITGB2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes residues 534-546 in cysteine-rich repeat 3 of the human CD18 (beta2 integrin). Cross-reacts with all three leukocyte integrins.

Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR