Anti-TNFSF15 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-859-C100
89155-160EA 539.55 USD
89155-160
Anti-TNFSF15 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For Immunoprecipitation, Western Blot

Recognizes human and mouse TL1A.

Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR