Anti-TNFSF15 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-855-C100
89155-150EA 558.08 USD
89155-150
Anti-TNFSF15 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Recognizes mouse TL1A.

Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR