Anti-CD3 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-822-C100
89155-054EA 334.63 USD
89155-054
Anti-CD3 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunoprecipitation

Recognizes the epsilon-chain of human CD3. Detects endogenous protein by FC, ICC and IP.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR