Anti-NALP3 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-819-C100
89155-048EA 462.16 USD
89155-048
Anti-NALP3 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human NLRP3/NALP3. Detects endogenous protein by IP and WB.

Type: Primary
Antigen: NALP3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR