Anti-APAF 1 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-348-C100
89154-082EA 477.42 USD
89154-082
Anti-APAF 1 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes the CARD domain of human Apaf-1. Does not cross-react with mouse or rat Apaf-1.

Type: Primary
Antigen: APAF 1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR