Anti-CTNNA1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-101-C100
89153-672EA 657.27 USD
89153-672
Anti-CTNNA1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and dog alpha-catenin. Does not cross-react with beta-catenins.

Reconstitute with 1ml distilled water (15 min., RT).

Type: Primary
Antigen: CTNNA1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR