Podophyllotoxin ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (10R,11R,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.03,7.011,15]hexadeca-1,3(7),8-trien-12-one, Condylox, PPT, (-)-Podophyllotoxin, Podofilox, Condyline, NSC 24818

630-086-M100
89152-968EA 56.68 USD
89152-968
Podophyllotoxin ≥98% (by HPLC)
Podophyllotoxin

Topoisomerase inhibitor


Potent inhibitor of microtubule assembly. Antineoplastic glucoside. Antitumor agent. DNA topoisomerase II inhibitor.


Isolated from Podophyllum peltatum.

Formula: C₂₂H₂₂O₈
MDL Number: MFCD00075290
CAS Number: 518-28-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403a42f4321f2e019d4f9ce4936bd4eb05472f86&vid=0d72a9a838b04fb7d81142173f75a9a8ea034fd9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR