DAF-FM diacetate 2,5 mg/ml ≥98% (by HPLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: DAF-FM DA, 4-Amino-5-methylamino-2',7'-difluorofluorescein diacetate

620-071-M001
89152-908EA 990.81 USD
89152-908
DAF-FM diacetate 2,5 mg/ml ≥98% (by HPLC)
DAF-FM diacetate

Fluorescent probe for NO detection. Cell permeable.


  • Formulation: Liquid.


Cell permeable photo-stable nitric oxide (NO) fluorescent indicator, that is hydrolyzed to DAF-FM by intracellular esterases (Ex: 500 nm; Em: 515 nm). Stable in a pH range of 5.5 – 12. High yield (quantum efficiency). Detection limit 3 nM.


Synonym: Diaminofluorescein-FM diacetate.

Formula: C₂₅H₁₈F₂N₂O₇
Storage Temperature: Freezer
CAS Number: 254109-22-3
EINECS: 200-664-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2748757cb0a457ab1217345748ed459fa499c1&vid=558c6157bca110b2e4712c669cba918d9e25b4bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation Dissolved in 0.4 mL DMSO
Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR