Caffeine ≥99%

Supplier: Enzo Life Sciences

550-322-G005
89152-518EA 50.14 USD
89152-518
Caffeine ≥99%
Caffeine

CNS stimulant


CNS stimulant. Blocks adenosine A1 and A2A receptors. cAMP phosphodiesterase inhibitor. Interferes with the uptake and storage of Ca2+ by the sarcoplasmic reticulum in skeletal muscle. Prevents apoptosis and cell cycle effects induced by various chemicals. Inhibits cellular DNA repair mechanisms. Anti-inflammatory.


Found in tea leaves, coffee beans, cocoa beans, maté leaves, guarana paste and kola nuts.

Formula: C₈H₁₀N₄O₂
MDL Number: MFCD00005758
CAS Number: 58-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae125620fb97282f202877db02e1edb9a728c22a&vid=25c9131ab458a7014b2270e6f44d3b149f172b82&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR