Pilocarpine hydrochloride ≥98% (by titrimetric analysis)

Supplier: Enzo Life Sciences

Synonyms: (+)-Pilocarpine hydrochloride

550-092-G001
89152-144EA 65.4 USD
89152-144
Pilocarpine hydrochloride ≥98% (by titrimetric analysis)
Pilocarpine hydrochloride

Cholinergic agonist


  • Formulation: White to off-white powder.


Cholinergic agonist.

Formula: C₁₁H₁₇ClN₂O₂
MDL Number: MFCD00012722
CAS Number: 54-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca3c24c6858fa52c825ecc9cc10db86bcff29e5f&vid=7979b3a1d67792155c48b256e09b60bf19de78ee&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR