5-Fluorouracil

Supplier: Enzo Life Sciences

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

480-099-G005
89151-608EA 103.55 USD
89151-608
5-Fluorouracil
5-Fluorouracil

Thymidylate synthetase inhibitor


Potent antitumor agent. Inhibits thymidylate synthetase. Induces p53-dependent apoptosis.

Formula: C₄H₃FN₂O₂
MDL Number: MFCD00006018
CAS Number: 51-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8aa090d6bc456753ec74dd83ebd9d04ef102cb9&vid=bd2893f864b8bca66a4efb6ef162d26b77cb2273&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR