Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt

480-021-G005
89151-498EA 77.39 USD
89151-498
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥98% (by HPLC)
Adenosine 5'-triphosphate disodium salt (ATP disodium salt)

  • Formulation: White to off-white powder.

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃
MW: 551.14 g/mol
MDL Number: MFCD00080339
CAS Number: 987-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4965f6b49c1175a212dfe16ff8c1e56e89b7d80&vid=c4d7131ac86736dd49f0fd062273b25808e5e3ed&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR