Vitamin B12 (cyanocobalamin) ≥96%

Supplier: Enzo Life Sciences

460-024-G001
89151-436EA 75.21 USD
89151-436 89151-438
Vitamin B12 (cyanocobalamin) ≥96%
Vitamin B12 (cyanocobalamin)

  • Formulation: Purple to red crystalline powder.


Synthetic.

Formula: C₆₃H₈₈CoN₁₄O₁₄P
MDL Number: MFCD00151092
CAS Number: 68-19-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55a29efc1d3b4aaf3e1123bfcd1db334d4ff0847&vid=fb1a37eb201b8301cc45d131d1141e9420ae6b34&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR