DL-α-Tocopherol (Vitamin E) ≥97%, high purity

Supplier: Enzo Life Sciences

Synonyms: (±)-α-Tocopherol, Vitamin E Alcohol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, 2,5,7,8-Tetramethyl-2-(4,8,12-trimethyltridecyl)chroman-6-ol, DL-alpha-Tocopherol, Vitamin E

460-018-G001
89151-428EA 49.05 USD
89151-428
DL-α-Tocopherol (Vitamin E) ≥97%, high purity
DL-α-Tocopherol

  • Formulation: Slightly yellow viscous oil.


Synthetic.

Formula: C₂₉H₅₀O₂
MW: 430.71 g/mol
MDL Number: MFCD00072051
CAS Number: 10191-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67238febf15be41f62307dc872e2e4f0f6f65986&vid=535aca0b72b5aac621f4b976f0794b65d9972a6b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR