Folic acid ≥97%

Supplier: Enzo Life Sciences

Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

460-006-G010
89151-408EA 51.23 USD
89151-408
Folic acid ≥97%
Folic acid

  • Formulation: Yellow to orange crystalline powder.

Formula: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
MDL Number: MFCD00079305
CAS Number: 59-30-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25180eccdd743a85e9a6aa85def1de2394f5fc05&vid=c01c4a7a25933b474c268cd06baf5704c0fe6dab&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR