Calcium pantothenate ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate

460-003-G010
89151-404EA 47.96 USD
89151-404
Calcium pantothenate ≥98% (by HPLC)
Calcium pantothenate

Precursor for coenzyme A biosynthesis .


  • Formulation: White to off-white powder.


Soluble in water. Also soluble in glycerol; slightly soluble in 100% ethanol or methanol; insoluble in acetone or ether.

Formula: C₁₈H₃₂CaN₂O₁₀
MW: 476.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002766
CAS Number: 137-08-6
EINECS: 205-278-9
Merck Index: 14,07015
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52ab6b2ec9bec1dcb6b6174160721c4b0cb67978&vid=e1b8ddacf4a8c5395d2b1da8a14a0c47bd563eee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) ≥98%
Identity Determined by IR
Appearance White to off-white powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR