Demecolcine ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
Colcemid
430-033-M001
430-033-M005
89150-938EA
55.59
USD
89150-938
89150-940
Demecolcine ≥98% (by HPLC)
Demecolcine
Depolymerizes microtubules and inhibits their formation. Induces apoptosis.
Semisynthetic from colchicine (Prod. No. ALX-380-033).
Formula:
C₂₁H₂₅NO₅ |
MDL Number:
MFCD00075459 CAS Number: 477-30-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...