DETA NONOate (NOC-18) ≥97% (by 1H-NMR)

Supplier: Enzo Life Sciences
Warning

Synonyms: DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene

430-014-5005 430-014-M005 430-014-M025
89150-876EA 206.01 USD
89150-876 89150-878 89150-880
DETA NONOate (NOC-18) ≥97% (by 1H-NMR)
DETA NONOate (NOC-18)

Nitric oxide (NO) donor.


  • Formulation: White to off-white solid.


Induces apoptosis in macrophages. The half-life time of DETA NONOate is 20 hours at 37 °C and 56 hours at 22 - 25 °C in 0.1 M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0. DETA NONOate liberates 2 mol of NO per mol of parent compound.


Soluble in water or ethanol. Relatively stable in alkaline solution.


Synonyms: DETA/NO, (Z)-1-[2-(2-aminoethyl)-N-(2-ammonioethyl)amino]diazen-1-ium-1,2-diolate, 3,3-bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene, 2,2'-(Hydroxynitrosohydrazino)bis-ethanamine


Caution: Protect from light and oxygen.

Formula: C₄H₁₃N₅O₂
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
MDL Number: MFCD00278808
CAS Number: 146724-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea3af089e35939dd81619730a13ca45bb888efc&vid=d527963469f1ebde6829f7bf3a22865566388c59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥97% (1H-NMR)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR