PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98% (by non aqueous titration)

Supplier: Enzo Life Sciences
Warning

430-007-M100
89150-846EA 51.3 USD
89150-846 89150-848
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98% (by non aqueous titration)
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl)

Stable nitric oxide (NO) radical scavenger.


  • Formulation: Blue crystals.


Soluble in methanol, 100% ethanol or water.


Synonym: 2-Phenyl-4,4,5,5-tetramethylimidazoline-1-oxyl-3-oxide

Formula: C₁₃H₁₇N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00059929
CAS Number: 18390-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d91d15f7a69857794c88ab339836b20c9f581829&vid=e612fce1800c54f57205f4524aefcb049570beee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (Nonaqueous Titration)
Purity Detail Purified by nonaqueous titration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR