Carboxy-PTIO potassium salt ≥99% (by HPLC)

Supplier: Enzo Life Sciences

430-001-M010 430-001-M050 430-001-M250
89150-824EA 75.21 USD
89150-824 89150-826 89150-828
Carboxy-PTIO potassium salt ≥99% (by HPLC)
Carboxy-PTIO potassium salt
Formula: C₁₄H₁₆KN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00216153
CAS Number: 148819-94-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00c60d89de66a0596a12a6402f961c29d8ec6c90&vid=047e0d3c0ef147e1721faadb055f12df861deebd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR