Sodium diethyldithiocarbamate trihydrate ≥99% (by titrimetric analysis)

Supplier: Enzo Life Sciences

400-003-G025
89150-570EA 77.39 USD
89150-570
Sodium diethyldithiocarbamate trihydrate ≥99% (by titrimetric analysis)
Sodium diethyldithiocarbamate trihydrate

Nitric oxide (NO) spin-trapping reagent. Thiol and iron chelator. Inhibits induction of macrophage nitric oxide synthase (NOS). Has been shown to be an inhibitor of the nuclear transcription factor κB (NF-κB).


  • NO spin trapping reagent


White to off-white solid.


Soluble in water (20mg/ml).

Formula: (C₂H₅)₂NCSSNa·3H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00150617
CAS Number: 20624-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03bd9cecdc024a921b0097f699408338daac46&vid=5cb4d83cd72e35bec6c52fe9241dca6742751150&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR