Myricetin ≥90% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one
385-012-M010
385-012-M050
89150-504EA
52.32
USD
89150-504
89150-506
Myricetin ≥90% (by HPLC)
Myricetin
Chemopreventive agent
Antioxidant flavonoid. Has antitumor and chemopreventive properties. Anti-inflammatory. Inhibits NF-kappaB activation.
PIM1 kinase inhibitor IC50=0.78µM.
Formula:
C₁₅H₁₀O₈ MW: 318.24 g/mol |
MDL Number:
MFCD00006827 CAS Number: 529-44-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...