Quercetin dihydrate ≥95% (by HPLC)

Supplier: Enzo Life Sciences

385-001-G005
89150-476EA 51.23 USD
89150-476 89150-478
Quercetin dihydrate ≥95% (by HPLC)
Quercetin dihydrate

Antioxidant flavonoid


Antioxidant flavonoid. Inhibitor of mitochondrial ATPase, cAMP- and cGMP-phosphodiesterases. Inhibitor of protein tyrosine kinases and protein kinase C (PKC). Induces apoptosis. Blocks cells at the G0/G1 interface. Activator of human deacetylase SIRT1. Reversible inhibitor of fatty acid synthase (FAS). Inhibits the production of the inflammatory mediators nitric oxide (NO), TNF-α and IL-12 in activated macrophages.Induces gastric relaxation in mouse stomach, mainly by suppressing Ca2+ influx through voltage-dependent calcium channels.


Isolated from Sophora japonica L.

Formula: C₁₅H₁₄O₉
MDL Number: MFCD00149487
CAS Number: 6151-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74d05f9065b6a4a975a8a7350ed71b2128c1fd76&vid=ca6cffd470f462102b94a3da78bc068ba23e36ae&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR