Erythromycin (from Streptomyces erythreus) ≥85% (dry basis)

Supplier: Enzo Life Sciences

380-274-G005
89150-328EA 78.48 USD
89150-328
Erythromycin (from Streptomyces erythreus) ≥85% (dry basis)
Erythromycin

Macrolide antibiotic


Inhibits protein synthesis (elongation) at the transpeptidation step. Binds to the 23S RNA of the 50S ribosomal subunit. Effective against Gram-negative and Gram-positive bacteria.


White to off-white powder


Soluble in water (50 mg/mL), 100% ethanol or methanol


Packaging: Shipping temperature: Ambient

Formula: C₃₇H₆₇NO₁₃
Storage Temperature: Freezer
MDL Number: MFCD00084654
CAS Number: 114-07-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=311ccd343956ae525a6297f56f616b5705e81cdf&vid=5fb19c082500af59d9d51d127fa5f7ec9cee4f4a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR