Blasticidin S hydrochloride (from Streptomyces griseochromogenes) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

380-089-M100
89149-988EA 507.94 USD
89149-988
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) ≥98% (by HPLC)
Blasticidin S hydrochloride

Antibiotic.


Inhibitor of DNA and protein synthesis.


White to off-white powder


Soluble in water, insoluble in organic solvents


Packaging: Shipping temperature: Ambient

Formula: C₁₇H₂₇ClN₈O₅
Storage Temperature: Refrigerator
MDL Number: MFCD02091640
CAS Number: 3513-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c3c534f791db2e1407169c4cc44c10d20c8a46b&vid=e37623a7dd2249d0ba01c2bb438d9b4b502b256b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR