Bafilomycin B1 (from Streptomyces griseus) ≥95% (by HPLC)

Supplier: Enzo Life Sciences

380-063-M001
89149-918EA 364.06 USD
89149-918
Bafilomycin B1 (from Streptomyces griseus) ≥95% (by HPLC)
Bafilomycin B1

V-ATPase inhibitor.


Macrolide antibiotic that acts as a specific inhibitor of vacuolar-type H+-ATPase.


Yellow solid


Slightly soluble in methanol or DMSO, insoluble in water


Packaging: Shipping temperature: Ambient

Formula: C₄₄H₆₅NO₁₃
Storage Temperature: Freezer
MDL Number: MFCD00242671
CAS Number: 88899-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=718ff1b1083806311a03d435fc37c8a6b399be1d&vid=471a3b826e794b76088f83fc104e5a14ca24dd61&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR