Hygromycin B (from Streptomyces hygroscopicus) ≥80% (by HPLC), liquid

Supplier: Enzo Life Sciences

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

380-059-UM01
89149-914EA 198.38 USD
89149-914
Hygromycin B (from Streptomyces hygroscopicus) ≥80% (by HPLC), liquid
Hygromycin B

Antibiotic and antiviral.


Antibiotic with high activity. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent.


Dark brown solution


Packaging: Shipping temperature: Ambient

Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06795479
CAS Number: 31282-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82f9034bb72c8de466ee01e6692a1abe4e2702e5&vid=60e04891368fd0781cb8a1962f2f70cd624603e0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR