Kanamycin sulfate (from Streptomyces kanamyceticus)

Supplier: Enzo Life Sciences

Synonyms: Kanamycin Monosulfate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulfate, Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

380-049-G005
89149-900EA 97.01 USD
89149-900
Kanamycin sulfate (from Streptomyces kanamyceticus)
Kanamycin sulphate

White to off-white crystalline powder


  • Contains 5% kanamycin B


Aminoglycoside antibiotic with broad antibacterial spectrum, including mycobacteria, many Gram-positive and most Gram-negative bacteria. Inhibits bacterial protein synthesis. Kanamycin resistance is widely used as a selection marker for obtaining transgenic plants.


Soluble in water


Packaging: Shipping temperature: Ambient

Formula: C₁₈H₃₈N₄O₁₅S
MW: 582.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00070253
CAS Number: 25389-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67fddcfe8d8cb8f931b1d510c16132edd5d563f1&vid=593a31cdc21a09ce9dacfe2d031023cd75c5e786&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR