Neomycin sulfate

Supplier: Enzo Life Sciences

Synonyms: Neomycin, Fradiomycin sulfate, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate, Fradiomycin sulphate, Neomycin sulphate salt, Neomycin sulfate salt, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulfate

380-035-G025
89149-866EA 515.57 USD
89149-866
Neomycin sulfate
Neomycin sulphate

Inhibitor of PLC and PLD.


Aminoglycoside antibiotic that inhibits translation by binding to the small subunit of prokaryotic ribosomes. Inhibitor of phospholipase C (PLC; IC50=1-10µM) due to binding to inositol phospholipids. Neomycin inhibits phosphatidylcholine-phospholipase D activity (IC50=65µM). Blocks voltage sensitive Ca2+ channels without affecting Na+/Ca2+ antiporters in nerve cells. Induces PLA2 activation in human platelets.

Formula: C₂₃H₄₆N₆O₁₃·3H₂SO₄
MW: 908.87 g/mol
MDL Number: MFCD00150827
CAS Number: 1405-10-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fcb6f917c70da9ecb3e56a495ee8f7e3afa46f46&vid=a3b5934d1c5247bca300fd824f4bbbfec5c691f8&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR