Colchicine ≥98% (by HPLC)

Supplier: Enzo Life Sciences

380-033-G001
89149-854EA 126.44 USD
89149-854
Colchicine ≥98% (by HPLC)
Colchicine

Inhibitor of tubulin polymerization


Depolymerizes microtubules and inhibits tubulin polymerization. Induces apoptosis in a variety of cells.


Isolated from Colchicum autumnale.

Formula: C₂₂H₂₅NO₆
MDL Number: MFCD00078484
CAS Number: 64-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=502321db5fd01618f8dbdcd931049e0f43eff8e4&vid=4f44f987a7109dd8906a956de058622b93c10466&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR