Streptozocin ≥97%

Supplier: Enzo Life Sciences

Synonyms: 1-Methyl-1-nitroso-3-((2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)urea, Streptozotocin

380-010-5100 380-010-G001 380-010-M100
89149-798EA 178.76 USD
89149-798 89149-800 89149-802
Streptozocin ≥97%
Streptozocin

Widely used diabetes inducer in rodents. Inhibition of β-cell O-GlcNAcase by streptozotocin is the mechanism that accounts for its diabetogenic toxicity. N-nitroso-containing antibiotic, acting as a nitric oxide (NO) donor and vasorelaxant. Relaxes phenylephrine-contracted aortic rings at 10µM. Potent methylating agent for DNA.


  • O-GlcNAcase inhibitor


White to pale yellow crystalline solid.


Soluble in water (nH2O at 102.8mg/ml) or 100% ethanol (200 proof at 0.92mg/ml).

Formula: C₈H₁₅N₃O₇
Storage Temperature: Freezer
MDL Number: MFCD00006607
CAS Number: 18883-66-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e301d071418a871ede32dbb2da5be6aa3a32f89&vid=30f0d26d00acba7b8b66f6043430b0baf481d9cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR