Hydroxytyrosol Acetate, Enzo Life Sciences
Supplier: Enzo Life Sciences
350-404-M050
89149-704EA 138.43 USD
89149-704
Hydroxytyrosol Acetate, Enzo Life Sciences
Cell Culture Supplements and Additives
Antioxidant
Synthetic
Antioxidant comparable to resveratrol (Prod. No. BML-FR104). Is converted to hydroxytyrosol through hydrolysis by esterases. Reaches full oxygen radical absorbance capacity (ORAC) when converted to free hydoxytyrosol. ORAC values of hydroxytyrosol are about 50% higher than resveratrol, 10-fold higher than L(+)-ascorbic acid and 4-fold higher than (-)-epicatechin. Has been shown to have anticancer and antiviral effects.Antibacterial at 400µg/mL against Streptococcus pyogenes, Staphylococcus aureus, Escherichia coli and Klebsiella pneumoniae.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e44faaecf51d343d96380ac292bbaa693ebbd0e&vid=7f588f0584c66a3af1169d0761b6e009698c36a8&prid=web&clid=SAPDAM