Resibufogenin (from Bufonis venom) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: (3β,5β,15β)-14,15-Epoxy-3-hydroxy-5-bufa-20,22-dienolide, 5-((1R,2aR,3aS,3bR,5aR,7S,9aS,9bS,11aR)-7-Hydroxy-9a,11a-dimethylhexadecahydronaphtho[1',2':6,7]indeno[1,7a-b]oxiren-1-yl)-2H-pyran-2-one

350-286-M010
89149-418EA 492.68 USD
89149-418
Resibufogenin (from Bufonis venom) ≥98% (by HPLC)
Resibufogenin

Specific Na+/K+-ATPase inhibitor.


  • Na+/K+ ATPase inhibitor


White to off-white solid.


Soluble in chloroform.

Formula: C₂₄H₃₂O₄
MW: 384.5 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00210361
CAS Number: 465-39-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99d502e9ecab379a26b4c7cd3822d9320e13855e&vid=6dd00bacf30c9cec4e6187549d31e01c2cfee5e1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR