Puerarin ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 8-(beta-D-Glucopyranosyl)-4',7-dihydroxyisoflavone

350-249-M005
89149-332EA 143.88 USD
89149-332
Puerarin ≥98% (by HPLC)
Puerarin

5-HT2c inhibitor


Biologically active isoflavone. Affects serotonin levels and platelet aggregation in blood cells. 5-HT2c antagonist. Antibacterial. Displays cardioprotective effects. Induces apoptosis.


Isolated from Kudzu root.

Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
MDL Number: MFCD00076007
CAS Number: 3681-99-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=580cfa8e51188a9324289bac481ac7830d686daa&vid=c56f0826eb44ae00b1a80067516646ce2fe74228&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR