Daidzin ≥98%

Supplier: Enzo Life Sciences

Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone, 3-(4-Hydroxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

350-248-M010
89149-328EA 127 USD
89149-328 89149-330
Daidzin ≥98%
Daidzin

Glucoside of daidzein.


  • Formulation: White to beige powder.


Glucoside of the isoflavone daidzein found in soy beans.


Soluble in DMSO or dilute aqueous base; insoluble in water.

Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00017466
CAS Number: 552-66-9
Merck Index: 14,02801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fddfa503153e5bf8f46fa66ce7c4bec8ea3d9c53&vid=4bdfb318e1289be110c65ac2d929e272610b0a43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR